Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 2 - 3 | 382 | 88 | |
| GenBank mRNA | AK067867.1 | - | 2 - 3 | 384 | 88 | |
| AK100614.1 | - | 1 - 2 | 73 | 88 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map