Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 5 - 6 | 774 | 104 | |
| GenBank mRNA | AK067867.1 | - | 5 - 6 | 1248 | 104 | |
| AK100614.1 | - | 4 - 5 | 465 | 104 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map