Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 8 - 9 | 1146 | 72 | |
| GenBank mRNA | AK067867.1 | - | 8 - 9 | 1620 | 72 | |
| AK100614.1 | - | 7 - 8 | 837 | 72 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map