Assay Details
| Gene Symbol: LOC4350402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761860.2 | XP_015617346.1 | 9 - 10 | 1196 | 132 | |
| GenBank mRNA | AK067867.1 | - | 9 - 10 | 1670 | 132 | |
| AK100614.1 | - | 8 - 9 | 887 | 132 | ||
More Information
Set Membership: |
|

LOC4350402
Genomic Map