Assay Details
| Gene Symbol: LOC4350686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759799.1 | XP_015615285.1 | 5 - 6 | 1448 | 104 | |
| XM_015759800.1 | XP_015615286.1 | 5 - 6 | 1461 | 104 | ||
| GenBank mRNA | AK064895.1 | - | 5 - 6 | 1410 | 104 | |
More Information
Set Membership: |
|

LOC4350686
Genomic Map