Assay Details
| Gene Symbol: LOC4350691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760134.2 | XP_015615620.1 | 1 - 2 | 356 | 122 | |
| GenBank mRNA | AK073687.1 | - | 1 - 2 | 357 | 122 | |
More Information
Set Membership: |
|

LOC4350691
Genomic Map