Assay Details
| Gene Symbol: LOC4350691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760134.2 | XP_015615620.1 | 8 - 9 | 858 | 72 | |
| GenBank mRNA | AK073687.1 | - | 8 - 9 | 859 | 72 | |
More Information
Set Membership: |
|

LOC4350691
Genomic Map