Assay Details
Gene Symbol: LOC4350714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015760088.2 | XP_015615574.1 | 12 - 13 | 1421 | 79 | |
GenBank mRNA | AK106372.1 | - | 6 - 7 | 493 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
