Assay Details
| Gene Symbol: LOC4351098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761101.2 | XP_015616587.1 | 5 - 6 | 987 | 100 | |
| GenBank mRNA | AK101124.1 | - | 5 - 6 | 1041 | 100 | |
More Information
Set Membership: |
|

LOC4351098
Genomic Map