Assay Details
| Gene Symbol: LOC4351098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761101.2 | XP_015616587.1 | 7 - 8 | 1140 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4351098| Gene Symbol: LOC4351098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761101.2 | XP_015616587.1 | 7 - 8 | 1140 | 64 | |
Set Membership: |
|