Assay Details
| Gene Symbol: LOC4325992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778198.2 | XP_015633684.1 | 1 - 2 | 540 | 58 | |
| XM_015778204.2 | XP_015633690.1 | 1 - 2 | 540 | 58 | ||
| GenBank mRNA | AK107424.1 | - | 1 - 2 | 308 | 58 | |
More Information
Set Membership: |
|

LOC4325992
Genomic Map