Assay Details
Gene Symbol: LOC4324413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765408.2 | XP_015620894.1 | 3 - 4 | 846 | 82 | |
GenBank mRNA | AK068747.1 | - | 3 - 4 | 823 | 82 | |
AK103737.1 | - | 3 - 4 | 805 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
