Assay Details
| Gene Symbol: LOC4327632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769105.2 | XP_015624591.1 | 7 - 8 | 945 | 121 | |
| XM_015769115.2 | XP_015624601.1 | 7 - 8 | 945 | 121 | ||
| XM_015769127.2 | XP_015624613.1 | 6 - 7 | 813 | 121 | ||
| XM_015769143.2 | XP_015624629.1 | 6 - 7 | 810 | 121 | ||
| GenBank mRNA | AK242074.1 | - | 2 - 3 | 282 | 121 | |
More Information
Set Membership: |
|

LOC4327632
Genomic Map