Assay Details
| Gene Symbol: LOC4326767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762239.2 | XP_015617725.1 | 5 - 6 | 1007 | 72 | |
| XM_015762248.2 | XP_015617734.1 | 6 - 7 | 924 | 72 | ||
| XM_015762257.2 | XP_015617743.1 | 6 - 7 | 908 | 72 | ||
| GenBank mRNA | AK243477.1 | - | 6 - 7 | 835 | 72 | |
More Information
Set Membership: |
|

LOC4326767
Genomic Map