Assay Details
| Gene Symbol: LOC4327785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766948.2 | XP_015622434.1 | 3 - 4 | 456 | 81 | |
| GenBank mRNA | AK064007.1 | - | 3 - 4 | 277 | 81 | |
| AK102521.1 | - | 3 - 4 | 310 | 81 | ||
| EU847008.1 | - | 3 - 4 | 291 | 81 | ||
More Information
Set Membership: |
|

LOC4327785
Genomic Map