Assay Details
| Gene Symbol: LOC4327785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766948.2 | XP_015622434.1 | 7 - 8 | 831 | 74 | |
| GenBank mRNA | AK064007.1 | - | 7 - 8 | 651 | 74 | |
| AK102521.1 | - | 7 - 8 | 685 | 74 | ||
| EU847008.1 | - | 7 - 8 | 666 | 74 | ||
More Information
Set Membership: |
|

LOC4327785
Genomic Map