Assay Details
| Gene Symbol: LOC4326362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760437.2 | XP_015615923.1 | 13 - 14 | 1155 | 101 | |
| GenBank mRNA | AK111838.1 | - | 12 - 13 | 1315 | 101 | |
More Information
Set Membership: |
|

LOC4326362
Genomic Map