Assay Details
| Gene Symbol: LOC4326227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767654.2 | XP_015623140.1 | 1 - 2 | 252 | 66 | |
| GenBank mRNA | AK100819.1 | - | 1 - 2 | 253 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015767654.2 transcripts.
Set Membership: |
|

LOC4326227
Genomic Map