Assay Details
| Gene Symbol: LOC4326227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767654.2 | XP_015623140.1 | 4 - 5 | 470 | 90 | |
| GenBank mRNA | AK100819.1 | - | 4 - 5 | 471 | 90 | |
More Information
Set Membership: |
|

LOC4326227
Genomic Map