Assay Details
| Gene Symbol: LOC4326227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767654.2 | XP_015623140.1 | 6 - 7 | 668 | 100 | |
| GenBank mRNA | AK100819.1 | - | 6 - 7 | 669 | 100 | |
More Information
Set Membership: |
|

LOC4326227
Genomic Map