Assay Details
| Gene Symbol: LOC9272440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766637.2 | XP_015622123.1 | 16 - 17 | 3056 | 134 | |
| GenBank mRNA | AK120868.1 | - | 15 - 16 | 4113 | 134 | |
More Information
Set Membership: |
|

LOC9272440
Genomic Map