Assay Details
| Gene Symbol: LOC9272440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766637.2 | XP_015622123.1 | 5 - 6 | 983 | 96 | |
| GenBank mRNA | AK120868.1 | - | 5 - 6 | 994 | 96 | |
More Information
Set Membership: |
|

LOC9272440
Genomic Map