Assay Details
| Gene Symbol: LOC9271759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766129.2 | XP_015621615.1 | 38 - 39 | 4606 | 149 | |
| GenBank mRNA | AK064446.1 | - | 1 - 2 | 310 | 149 | |
More Information
Set Membership: |
|

LOC9271759
Genomic Map