Assay Details
| Gene Symbol: LOC4325847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759688.2 | XP_015615174.1 | 1 - 2 | 1044 | 104 | |
| XM_015759692.2 | XP_015615178.1 | 1 - 2 | 1045 | 104 | ||
| XM_015759699.2 | XP_015615185.1 | 1 - 2 | 1047 | 104 | ||
| XM_015759707.2 | XP_015615193.1 | 1 - 2 | 1048 | 104 | ||
| XM_015759715.2 | XP_015615201.1 | 1 - 2 | 1048 | 104 | ||
| GenBank mRNA | AK243249.1 | - | 1 - 2 | 327 | 104 | |
More Information
Set Membership: |
|

LOC4325847
Genomic Map