Assay Details
| Gene Symbol: LOC4325847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759688.2 | XP_015615174.1 | 3 - 4 | 1278 | 72 | |
| XM_015759692.2 | XP_015615178.1 | 3 - 4 | 1279 | 72 | ||
| XM_015759699.2 | XP_015615185.1 | 3 - 4 | 1281 | 72 | ||
| XM_015759707.2 | XP_015615193.1 | 3 - 4 | 1282 | 72 | ||
| XM_015759715.2 | XP_015615201.1 | 3 - 4 | 1282 | 72 | ||
| GenBank mRNA | AK243249.1 | - | 3 - 4 | 561 | 72 | |
More Information
Set Membership: |
|

LOC4325847
Genomic Map