Assay Details
| Gene Symbol: LOC4325847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759688.2 | XP_015615174.1 | 4 - 5 | 1372 | 114 | |
| XM_015759692.2 | XP_015615178.1 | 4 - 5 | 1373 | 114 | ||
| XM_015759699.2 | XP_015615185.1 | 4 - 5 | 1375 | 114 | ||
| XM_015759715.2 | XP_015615201.1 | 4 - 5 | 1376 | 114 | ||
| GenBank mRNA | AK243249.1 | - | 4 - 5 | 655 | 114 | |
More Information
Set Membership: |
|

LOC4325847
Genomic Map