Assay Details
| Gene Symbol: LOC4326314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762322.2 | XP_015617808.1 | 11 - 12 | 1002 | 150 | |
| GenBank mRNA | AK070726.1 | - | 11 - 12 | 1012 | 150 | |
| AK101558.1 | - | 3 - 4 | 273 | 150 | ||
| AK101859.1 | - | 11 - 12 | 969 | 150 | ||
More Information
Set Membership: |
|

LOC4326314
Genomic Map