Assay Details
| Gene Symbol: LOC4326314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762322.2 | XP_015617808.1 | 13 - 14 | 1162 | 100 | |
| GenBank mRNA | AK070726.1 | - | 13 - 14 | 1172 | 100 | |
| AK101558.1 | - | 5 - 6 | 433 | 100 | ||
| AK101859.1 | - | 13 - 14 | 1129 | 100 | ||
More Information
Set Membership: |
|

LOC4326314
Genomic Map