Assay Details
| Gene Symbol: LOC4326314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762322.2 | XP_015617808.1 | 16 - 17 | 1536 | 113 | |
| GenBank mRNA | AK070726.1 | - | 16 - 17 | 1547 | 114 | |
| AK101558.1 | - | 8 - 9 | 807 | 113 | ||
| AK101859.1 | - | 16 - 17 | 1504 | 114 | ||
More Information
Set Membership: |
|

LOC4326314
Genomic Map