Assay Details
| Gene Symbol: LOC4326314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762322.2 | XP_015617808.1 | 1 - 2 | 162 | 62 | |
| GenBank mRNA | AK070726.1 | - | 1 - 2 | 172 | 62 | |
| AK101859.1 | - | 1 - 2 | 129 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015762322.2 transcripts.
Set Membership: |
|

LOC4326314
Genomic Map