Assay Details
| Gene Symbol: LOC4326314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762322.2 | XP_015617808.1 | 20 - 21 | 2163 | 91 | |
| GenBank mRNA | AK101558.1 | - | 12 - 13 | 1434 | 91 | |
| AK101859.1 | - | 20 - 21 | 2131 | 91 | ||
More Information
Set Membership: |
|

LOC4326314
Genomic Map