Assay Details
| Gene Symbol: LOC4327659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755112.1 | XP_015610598.1 | 4 - 5 | 1098 | 75 | |
| XM_015755759.1 | XP_015611245.1 | 4 - 5 | 1045 | 75 | ||
| GenBank mRNA | AK069698.1 | - | 2 - 3 | 1778 | 75 | |
More Information
Set Membership: |
|

LOC4327659
Genomic Map