Assay Details
| Gene Symbol: LOC4327659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755112.1 | XP_015610598.1 | 5 - 6 | 1282 | 102 | |
| XM_015755759.1 | XP_015611245.1 | 5 - 6 | 1229 | 102 | ||
| GenBank mRNA | AK069698.1 | - | 3 - 4 | 1962 | 102 | |
More Information
Set Membership: |
|

LOC4327659
Genomic Map