Assay Details
| Gene Symbol: LOC4326617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766409.2 | XP_015621895.1 | 11 - 12 | 1589 | 82 | |
| GenBank mRNA | AK068431.1 | - | 2 - 3 | 91 | 82 | |
More Information
Set Membership: |
|

LOC4326617
Genomic Map