Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 11 - 12 | 1170 | 81 | |
| XM_015757449.2 | XP_015612935.1 | 11 - 12 | 1033 | 81 | ||
| GenBank mRNA | AB110983.1 | - | 11 - 12 | 824 | 81 | |
| AK102997.1 | - | 11 - 12 | 1009 | 81 | ||
More Information
Set Membership: |
|

LOC4324139
Genomic Map