Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 12 - 13 | 1278 | 80 | |
| XM_015757449.2 | XP_015612935.1 | 12 - 13 | 1141 | 80 | ||
| GenBank mRNA | AK102997.1 | - | 12 - 13 | 1117 | 80 | |
More Information
Set Membership: |
|

LOC4324139
Genomic Map