Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 1 - 2 | 327 | 125 | |
| XM_015757449.2 | XP_015612935.1 | 1 - 2 | 325 | 125 | ||
| GenBank mRNA | AB110983.1 | - | 1 - 2 | 116 | 125 | |
| AK102997.1 | - | 1 - 2 | 166 | 125 | ||
More Information
Set Membership: |
|

LOC4324139
Genomic Map