Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 2 - 3 | 560 | 111 | |
| GenBank mRNA | AK102997.1 | - | 2 - 3 | 399 | 111 | |
More Information
Set Membership: |
|

LOC4324139
Genomic Map