Assay Details
| Gene Symbol: LOC4324139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757442.2 | XP_015612928.1 | 6 - 7 | 762 | 107 | |
| XM_015757449.2 | XP_015612935.1 | 6 - 7 | 625 | 107 | ||
| GenBank mRNA | AB110983.1 | - | 6 - 7 | 416 | 107 | |
| AK102997.1 | - | 6 - 7 | 601 | 107 | ||
More Information
Set Membership: |
|

LOC4324139
Genomic Map