Assay Details
| Gene Symbol: LOC4327939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766340.2 | XP_015621826.1 | 5 - 6 | 785 | 63 | |
| GenBank mRNA | AK120982.1 | - | 5 - 6 | 714 | 63 | |
More Information
Set Membership: |
|

LOC4327939
Genomic Map