Assay Details
| Gene Symbol: LOC4325617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773120.2 | XP_015628606.1 | 2 | 580 | 91 | |
| GenBank mRNA | AK120226.1 | - | 1 | 757 | 91 | |
More Information
Set Membership: |
|

LOC4325617
Genomic Map