Assay Details
| Gene Symbol: LOC4326892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766354.2 | XP_015621840.1 | 1 - 2 | 250 | 83 | |
| XM_015766356.2 | XP_015621842.1 | 1 - 2 | 250 | 83 | ||
| GenBank mRNA | AK071863.1 | - | 1 - 2 | 231 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015766354.2, XM_015766356.2 transcripts.
Set Membership: |
|

LOC4326892
Genomic Map