Assay Details
| Gene Symbol: LOC4326892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766354.2 | XP_015621840.1 | 3 - 4 | 478 | 64 | |
| XM_015766355.2 | XP_015621841.1 | 3 - 4 | 570 | 64 | ||
| XM_015766356.2 | XP_015621842.1 | 3 - 4 | 478 | 64 | ||
| GenBank mRNA | AK071863.1 | - | 3 - 4 | 459 | 64 | |
More Information
Set Membership: |
|

LOC4326892
Genomic Map