Assay Details
| Gene Symbol: LOC4326892 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766356.2 | XP_015621842.1 | 5 - 6 | 651 | 158 | |
| GenBank mRNA | AK071863.1 | - | 5 - 6 | 632 | 158 | |
More Information
Set Membership: |
|

LOC4326892
Genomic Map