Assay Details
| Gene Symbol: LOC4327841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243093.1 | - | 2 | 4304 | 109 | |
| AY044444.1 | - | 1 | 4453 | 109 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4327841| Gene Symbol: LOC4327841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK243093.1 | - | 2 | 4304 | 109 | |
| AY044444.1 | - | 1 | 4453 | 109 | ||
Set Membership: |
|