Assay Details
| Gene Symbol: LOC4327483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765228.2 | XP_015620714.1 | 1 - 2 | 102 | 90 | |
| GenBank mRNA | AK058612.1 | - | 1 - 2 | 52 | 90 | |
| AK101927.1 | - | 1 - 2 | 53 | 90 | ||
| BT131988.1 | - | 1 - 2 | 53 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015765228.2 transcripts.
Set Membership: |
|

LOC4327483
Genomic Map