Assay Details
| Gene Symbol: LOC4327483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765228.2 | XP_015620714.1 | 5 - 6 | 470 | 83 | |
| GenBank mRNA | AB118010.1 | - | 1 - 2 | 78 | 83 | |
| AK058612.1 | - | 5 - 6 | 420 | 83 | ||
| AK101927.1 | - | 5 - 6 | 421 | 83 | ||
| BT131058.1 | - | 1 - 2 | 129 | 83 | ||
| BT131988.1 | - | 5 - 6 | 421 | 83 | ||
| Gene Symbol: LOC4323935 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT132583.1 | - | 112 | 83 | ||
More Information
Set Membership: |
|

LOC4327483
Genomic Map