Assay Details
| Gene Symbol: LOC4327469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762710.2 | XP_015618196.1 | 8 - 9 | 1470 | 69 | |
| XM_015762717.2 | XP_015618203.1 | 8 - 9 | 1463 | 69 | ||
| GenBank mRNA | AK063080.1 | - | 3 - 4 | 308 | 69 | |
More Information
Set Membership: |
|

LOC4327469
Genomic Map