Assay Details
| Gene Symbol: LOC4327469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762710.2 | XP_015618196.1 | 10 - 11 | 1594 | 126 | |
| XM_015762717.2 | XP_015618203.1 | 10 - 11 | 1587 | 126 | ||
| GenBank mRNA | AK063080.1 | - | 5 - 6 | 432 | 126 | |
More Information
Set Membership: |
|

LOC4327469
Genomic Map