Assay Details
| Gene Symbol: LOC4324939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785781.2 | XP_015641267.1 | 11 - 12 | 1313 | 102 | |
| XM_015785787.2 | XP_015641273.1 | 11 - 12 | 1307 | 102 | ||
| XM_015785797.1 | XP_015641283.1 | 12 - 13 | 1318 | 102 | ||
| XM_015785805.1 | XP_015641291.1 | 12 - 13 | 1312 | 102 | ||
| GenBank mRNA | AK064743.1 | - | 11 - 12 | 1301 | 102 | |
More Information
Set Membership: |
|

LOC4324939
Genomic Map